Assay Details
Target Gene Details
Entrez Gene ID: | 285819 |
Gene Name: | uncharacterized LOC285819 |
Gene Aliases: |
- |
Location: |
Chr.6:26471944-26482509 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC285819 | NR_038992.1 | 3 | 2007 | |
| AK097539.1 | 3 | 2007 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023453 | Chr.6:25853602 - 26528022 on Build GRCh38 | Gain |
|
| nsv1031938 | Chr.6:26466954 - 26501669 on Build GRCh38 | Loss |
|

Genomic Map