Assay Details
Target Gene Details
Entrez Gene ID: | 57706 |
Gene Name: | DENN domain containing 1A |
Gene Aliases: |
FAM31A, KIAA1608 |
Location: |
Chr.9:123379654-123930158 on Build GRCh38 |
Assay Gene Location: | Within Intron 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DENND1A | NM_020946.1 | NP_065997.1 | ||
| XM_005252109.2 | XP_005252166.1 | |||
| XM_005252111.4 | XP_005252168.1 | |||
| XM_006717195.3 | XP_006717258.1 | |||
| XM_011518882.2 | XP_011517184.1 | |||
| XM_011518883.2 | XP_011517185.1 | |||
| XM_011518884.2 | XP_011517186.1 | |||
| XM_011518885.2 | XP_011517187.1 | |||
| XM_011518886.2 | XP_011517188.1 | |||
| XM_017014947.1 | XP_016870436.1 | |||
| XM_017014948.1 | XP_016870437.1 | |||
| XM_017014949.1 | XP_016870438.1 | |||
| XM_017014950.1 | XP_016870439.1 | |||
| XM_017014951.1 | XP_016870440.1 | |||
| XM_017014952.1 | XP_016870441.1 | |||
| AB046828.2 | BAB13434.2 | |||
| AK074151.1 | BAB84977.1 | |||
| AK295710.1 | ||||
| AK299867.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv615285 | Chr.9:123365719 - 123425419 on Build GRCh38 | Loss |
|
| nsv518647 | Chr.9:123287696 - 123433925 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map