Assay Details
Target Gene Details
Entrez Gene ID: | 4048 |
Gene Name: | leukotriene A4 hydrolase |
Gene Aliases: |
- |
Location: |
Chr.12:96000753-96043520 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LTA4H | NM_000895.2 | NP_000886.1 | ||
| NM_001256643.1 | NP_001243572.1 | |||
| NM_001256644.1 | NP_001243573.1 | |||
| NR_132659.1 | ||||
| XM_005268871.1 | XP_005268928.1 | |||
| XM_011538348.1 | XP_011536650.1 | |||
| AB209740.1 | BAD92977.1 | |||
| AK293630.1 | ||||
| AK298017.1 | ||||
| AK301308.1 | ||||
| BC032528.1 | AAH32528.1 | |||
| BC036555.1 | AAH36555.1 | |||
| BX647158.1 | ||||
| CR457068.1 | CAG33349.1 | |||
| J02959.1 | AAA36177.1 | |||
| J03459.1 | AAA36176.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3630445 | Chr.12:95990852 - 96016216 on Build GRCh38 | Gain |
|
| nsv523292 | Chr.12:95992360 - 96015423 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map