Assay Details
Target Gene Details
Entrez Gene ID: | 121506 |
Gene Name: | endoplasmic reticulum protein 27 |
Gene Aliases: |
C12orf46, PDIA8 |
Location: |
Chr.12:14914027-14938549 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERP27 | NM_152321.3 | NP_689534.1 | ||
| AK056677.1 | BAB71251.1 | |||
| AY358536.1 | AAQ88900.1 | |||
| BC030218.1 | AAH30218.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469120 | Chr.12:14922268 - 14948501 on Build GRCh38 | Gain |
|
| nsv557612 | Chr.12:14921379 - 14939445 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map