Assay Details
Target Gene Details
Entrez Gene ID: | 283455 |
Gene Name: | kinase suppressor of ras 2 |
Gene Aliases: |
- |
Location: |
Chr.12:117453012-117968558 on Build GRCh38 |
Assay Gene Location: | Within Intron 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KSR2 | NM_173598.4 | NP_775869.3 | ||
| XM_011538224.2 | XP_011536526.1 | |||
| XM_011538225.2 | XP_011536527.1 | |||
| XM_011538228.2 | XP_011536530.1 | |||
| XM_017019208.1 | XP_016874697.1 | |||
| XM_017019210.1 | XP_016874699.1 | |||
| AY345972.1 | AAQ24226.1 | |||
| BC107106.1 | AAI07107.1 | |||
| BC107107.1 | AAI07108.1 | |||
| BC127603.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560386 | Chr.12:117384382 - 117553742 on Build GRCh38 | Gain |
|
| esv3581916 | Chr.12:117356518 - 118228514 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map