Assay Details
Target Gene Details
Entrez Gene ID: | 144348 |
Gene Name: | zinc finger protein 664 |
Gene Aliases: |
ZFOC1, ZNF176 |
Location: |
Chr.12:123973215-124015439 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF664 | NM_001204298.1 | NP_001191227.1 | ||
| NM_152437.2 | NP_689650.1 | |||
| AK056034.1 | ||||
| AL834266.1 | CAD38941.1 | |||
| BC051696.1 | AAH51696.1 | |||
| BX647762.1 | CAH10567.1 | |||
| DA304849.1 | ||||
| DA328413.1 |
Target Gene Details
Entrez Gene ID: | 100533183 |
Gene Name: | filamin-interacting protein FAM101A |
Gene Aliases: |
- |
Location: |
Chr.12:123973215-124316024 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF664-FAM101A | NM_001204299.1 | NP_001191228.1 | ||
| DA169732.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2746515 | Chr.12:123974024 - 124011545 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map