Assay Details
Target Gene Details
Entrez Gene ID: | 11100 |
Gene Name: | heterogeneous nuclear ribonucleoprotein U like 1 |
Gene Aliases: |
E1B-AP5, E1BAP5, HNRPUL1 |
Location: |
Chr.19:41262476-41307783 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPUL1 | NM_144732.4 | NP_653333.1 | ||
| XM_005258464.3 | XP_005258521.1 | |||
| XM_011526393.2 | XP_011524695.1 | |||
| XM_011526395.2 | XP_011524697.1 | |||
| AK023006.1 | ||||
| AK225850.1 | ||||
| AK308078.1 | ||||
| DC347248.1 | ||||
| DC366321.1 |
More Information
Set Membership: |
|

Genomic Map