Assay Details
Target Gene Details
Entrez Gene ID: | 10653 |
Gene Name: | serine peptidase inhibitor, Kunitz type 2 |
Gene Aliases: |
DIAR3, HAI-2, HAI2, Kop, PB |
Location: |
Chr.19:38264458-38292614 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPINT2 | NM_001166103.1 | NP_001159575.1 | ||
| NM_021102.3 | NP_066925.1 | |||
| AB006534.1 | BAA25024.1 | |||
| AF027205.1 | AAB84031.1 | |||
| AK291532.1 | ||||
| AK297154.1 | ||||
| AX772830.1 | CAE06264.1 | |||
| BC001668.2 | AAH01668.1 | |||
| BC007705.2 | AAH07705.1 | |||
| BC011951.1 | AAH11951.1 | |||
| BC011955.1 | AAH11955.1 | |||
| BC012868.2 | AAH12868.1 | |||
| BT020115.1 | AAV38918.1 | |||
| BT020116.1 | AAV38919.1 | |||
| BT020117.1 | AAV38920.1 | |||
| CR407604.1 | CAG28532.1 | |||
| U78095.1 | AAC02781.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833822 | Chr.19:38270108 - 38454465 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map