Assay Details
Target Gene Details
Entrez Gene ID: | 57325 |
Gene Name: | lysine acetyltransferase 14 |
Gene Aliases: |
ATAC2, CRP2BP, CSRP2BP, PRO1194, dJ717M23.1 |
Location: |
Chr.20:18142224-18188387 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KAT14 | NM_020536.4 | NP_065397.3 | ||
| BC007537.1 | AAH07537.1 | |||
| BQ691686.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv585665 | Chr.20:18106305 - 18144797 on Build GRCh38 | Gain |
|
| nsv833933 | Chr.20:18029399 - 18206602 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map