Assay Details
Target Gene Details
Entrez Gene ID: | 128710 |
Gene Name: | SLX4 interacting protein |
Gene Aliases: |
C20orf94, bA204H22.1, bA254M13.1, dJ1099D15.3 |
Location: |
Chr.20:10434453-10628138 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLX4IP | NM_001009608.2 | NP_001009608.1 | ||
| XM_017027648.1 | XP_016883137.1 | |||
| XM_017027650.1 | XP_016883139.1 | |||
| XM_017027652.1 | XP_016883141.1 | |||
| BC020787.1 | ||||
| BC026094.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522874 | Chr.20:9918791 - 10838562 on Build GRCh38 | Loss |
|
| nsv833920 | Chr.20:10460192 - 10651051 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map