Assay Details
Target Gene Details
Entrez Gene ID: | 11237 |
Gene Name: | ring finger protein 24 |
Gene Aliases: |
G1L |
Location: |
Chr.20:3927309-4015591 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF24 | NM_001134337.2 | NP_001127809.1 | ||
| NM_001134338.2 | NP_001127810.1 | |||
| NM_001321749.1 | NP_001308678.1 | |||
| NM_007219.4 | NP_009150.1 | |||
| XM_011529145.2 | XP_011527447.1 | |||
| XM_011529146.2 | XP_011527448.1 | |||
| XM_011529147.2 | XP_011527449.1 | |||
| XM_017027620.1 | XP_016883109.1 | |||
| XM_017027621.1 | XP_016883110.1 | |||
| XM_017027622.1 | XP_016883111.1 | |||
| AL079313.1 | CAB45279.1 | |||
| AL096778.1 | CAB46627.1 | |||
| BC000213.2 | AAH00213.1 | |||
| BC039584.1 | AAH39584.1 | |||
| BT007406.1 | AAP36074.1 | |||
| CV811405.1 | ||||
| HY140488.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3645053 | Chr.20:3849918 - 3987827 on Build GRCh38 | Loss |
|
| nsv828673 | Chr.20:3738382 - 4184811 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76354766] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map