Assay Details
Target Gene Details
Entrez Gene ID: | 100303755 |
Gene Name: | PET117 homolog |
Gene Aliases: |
CSRP2BP |
Location: |
Chr.20:18137855-18143169 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PET117 | NM_001164811.1 | NP_001158283.1 | ||
| AY358666.1 | AAQ89029.1 | |||
| BI601826.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv585665 | Chr.20:18106305 - 18144797 on Build GRCh38 | Gain |
|
| nsv833933 | Chr.20:18029399 - 18206602 on Build GRCh38 | Gain |
|

Genomic Map