Assay Details
Target Gene Details
Entrez Gene ID: | 100528031 |
Gene Name: | FKBP1A-SDCBP2 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.20:1309909-1393172 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FKBP1A-SDCBP2 | NR_037661.1 | |||
| AK309026.1 | ||||
| BM925547.1 |
Target Gene Details
Entrez Gene ID: | 100507495 |
Gene Name: | SDCBP2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.20:1325343-1378735 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDCBP2-AS1 | NR_040047.1 | |||
| NR_040048.1 | ||||
| NR_040049.1 | ||||
| AK055993.1 | ||||
| DA274215.1 | ||||
| DB454162.1 |

Genomic Map