Assay Details
Target Gene Details
Entrez Gene ID: | 4666 |
Gene Name: | nascent polypeptide-associated complex alpha subunit |
Gene Aliases: |
HSD48, NAC-alpha, NACA1, skNAC |
Location: |
Chr.12:56712427-56726275 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
| esv22791 | Chr.12:56716780 - 56721722 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map