Assay Details
Target Gene Details
Entrez Gene ID: | 71446 |
Gene Name: | tryptophan rich basic protein |
Gene Aliases: |
5530402J05Rik, C030018G21Rik, Chd5 |
Location: |
Chr.16:96145354-96189247 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wrb | NM_207301.2 | NP_997184.1 | ||
| AK021091.1 | 4 | 523 | ||
| AK172079.1 | BAE42815.1 | |||
| BC031769.1 | AAH31769.1 |

Genomic Map