Assay Details
Target Gene Details
Entrez Gene ID: | 208718 |
Gene Name: | DIS3 mitotic control homolog (S. cerevisiae)-like 2 |
Gene Aliases: |
4930429A22Rik, 8030493P09Rik |
Location: |
Chr.1:86703779-87050097 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dis3l2 | NM_001172157.1 | NP_001165628.1 | ||
| NM_153530.2 | NP_705758.1 | |||
| AK029652.1 | BAC26549.1 | |||
| AK031180.1 | BAC27292.1 | |||
| AK040613.1 | ||||
| AK213947.1 | ||||
| AK214096.1 | ||||
| BC036177.1 | AAH36177.1 |
More Information
Set Membership: |
|

Genomic Map