Assay Details
Target Gene Details
Entrez Gene ID: | 210356 |
Gene Name: | NCK-associated protein 5 |
Gene Aliases: |
8430408F21, D130011D22Rik, E030049G20Rik, Erih1, Erih2, Gm1548, Nap5 |
Location: |
Chr.1:125913636-126830897 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nckap5 | NM_001081756.1 | NP_001075225.1 | ||
| NM_176957.3 | NP_795931.2 | |||
| AK041950.1 | BAC31107.1 | |||
| AK083792.1 | ||||
| CN700744.1 |

Genomic Map