Assay Details
Target Gene Details
Entrez Gene ID: | 100039138 |
Gene Name: | predicted gene 2061 |
Gene Aliases: |
- |
Location: |
Chr.1:185447116-185455568 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm2061 | NR_040305.1 | |||
| AK138296.1 |
More Information
Set Membership: |
|

Genomic Map