Assay Details
Target Gene Details
Entrez Gene ID: | 14194 |
Gene Name: | fumarate hydratase 1 |
Gene Aliases: |
Fh, Fh-1 |
Location: |
Chr.1:175601378-175625635 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fh1 | NM_010209.2 | NP_034339.2 | ||
| AK002379.1 | ||||
| AK147000.1 | BAE27597.1 | |||
| AK194387.1 | ||||
| AK213605.1 | ||||
| BC006048.1 | AAH06048.1 | |||
| BU526396.1 | ||||
| U72679.1 | AAB51039.1 |
More Information
Set Membership: |
|

Genomic Map