Assay Details
Target Gene Details
Entrez Gene ID: | 226517 |
Gene Name: | Smg-7 homolog, nonsense mediated mRNA decay factor (C. elegans) |
Gene Aliases: |
9430023P16Rik, mKIAA0250 |
Location: |
Chr.1:152836995-152902686 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smg7 | NM_001005507.2 | 15 | 2020 | NP_001005507.1 |
| NM_001160256.1 | 15 | 1882 | NP_001153728.1 | |
| NM_001160257.1 | 15 | 1872 | NP_001153729.1 | |
| AK129101.1 | 15 | 1872 | BAC97911.1 | |
| BC082789.1 | 15 | 2007 | AAH82789.1 | |
| BC086651.1 | 15 | 1882 | AAH86651.1 |
More Information
Set Membership: |
|

Genomic Map