Assay Details
Target Gene Details
Entrez Gene ID: | 329540 |
Gene Name: | nucleolar protein 4-like |
Gene Aliases: |
4931410H15, 8430427H17Rik, AI429157, Gm1636 |
Location: |
Chr.2:153407461-153529971 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nol4l | NM_001134300.2 | 3 | 746 | NP_001127772.1 |
| AK029844.1 | 5 | 1225 | ||
| AK147524.1 | 3 | 748 | BAE27972.1 | |
| BC075648.1 | 2 | 157 | AAH75648.1 |

Genomic Map