Assay Details
Target Gene Details
Entrez Gene ID: | 74192 |
Gene Name: | actin related protein 2/3 complex, subunit 5-like |
Gene Aliases: |
2010015J01Rik, AI852867, ARC16-2, AW555592, AW742746 |
Location: |
Chr.2:39005272-39015878 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arpc5l | NM_028809.1 | 1 | 152 | NP_083085.1 |
| AK008255.1 | 1 | 152 | BAB25561.1 | |
| BC024482.1 | 1 | 175 | AAH24482.1 |
More Information
Set Membership: |
|

Genomic Map