Assay Details
Target Gene Details
Entrez Gene ID: | 228094 |
Gene Name: | ceramide kinase-like |
Gene Aliases: |
Gm1958, Rp26 |
Location: |
Chr.2:79331105-79430520 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cerkl | NM_001048176.1 | 12 | 1548 | NP_001041641.1 |
| CJ235603.1 | 6 | 479 | ||
| HQ426680.1 | 12 | 1548 | ||
| HQ426681.1 | 10 | 1341 | ||
| HQ426682.1 | 9 | 1209 | ||
| HQ426683.1 | 8 | 966 | ||
| HQ426684.1 | 11 | 1416 | ||
| HQ426685.1 | 11 | 1488 | ||
| HQ426686.1 | 11 | 1405 | ||
| HQ426687.1 | 12 | 1679 | ||
| HQ426688.1 | 12 | 1695 | ||
| HQ426689.1 | 11 | 1563 | ||
| HQ426690.1 | 12 | 1765 | ||
| HQ426702.1 | 11 | 1130 | ||
| HQ426703.1 | 9 | 923 | ||
| HQ426704.1 | 12 | 1287 | ||
| HQ426705.1 | 10 | 987 | ||
| HQ426706.1 | 10 | 1074 | ||
| HQ426707.1 | 10 | 1221 | ||
| HQ426708.1 | 6 | 584 | ||
| HQ426709.1 | 9 | 979 | ||
| HQ426710.1 | 8 | 919 | ||
| HQ426711.1 | 9 | 1126 |
More Information
Set Membership: |
|

Genomic Map