Assay Details
Target Gene Details
Entrez Gene ID: | 19025 |
Gene Name: | cathepsin A |
Gene Aliases: |
AU019505, PPCA, Ppgb |
Location: |
Chr.2:164832872-164841032 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctsa | NM_001038492.2 | NP_001033581.1 | ||
| NM_008906.4 | NP_032932.2 | |||
| AK019861.1 | BAB31888.1 | |||
| AK032197.1 | BAC27752.1 | |||
| AK159150.1 | BAE34857.1 | |||
| AK159766.1 | BAE35356.1 | |||
| AK179070.1 | ||||
| AK184500.1 | ||||
| BC018534.1 | AAH18534.1 | |||
| J05261.1 | AAA39982.1 |
More Information
Set Membership: |
|

Genomic Map