Assay Details
Target Gene Details
Entrez Gene ID: | 228714 |
Gene Name: | cysteine and glycine-rich protein 2 binding protein |
Gene Aliases: |
2510008M08Rik, ATAC2, AU023459, D2Ertd473e, D2Wsu131e, E430020F17 |
Location: |
Chr.2:144369032-144407675 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csrp2bp | NM_001166640.1 | NP_001160112.1 | ||
| NM_181417.3 | NP_852082.2 | |||
| BC031563.1 | AAH31563.1 |
Target Gene Details
Entrez Gene ID: | 100048644 |
Gene Name: | PET117 homolog (S. cerevisiae) |
Gene Aliases: |
Gm20571 |
Location: |
Chr.2:144368983-144373337 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pet117 | NM_001164813.1 | NP_001158285.1 |

Genomic Map