Assay Details
Target Gene Details
Entrez Gene ID: | 80744 |
Gene Name: | CWC22 spliceosome-associated protein homolog (S. cerevisiae) |
Gene Aliases: |
AA684037, AI173004, AL022752, B230213M24, mKIAA1604 |
Location: |
Chr.2:77895653-77946356 on Build GRCm38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cwc22 | NM_001290740.1 | NP_001277669.1 | ||
| NM_030560.5 | NP_085037.2 | |||
| NM_172667.2 | NP_766255.1 | |||
| AK045589.1 | BAC32427.1 | |||
| AK077961.1 | BAC37085.1 | |||
| AK132001.1 | BAE20931.1 | |||
| AK149528.1 | BAE28941.1 | |||
| AK166487.1 | BAE38804.1 | |||
| BC003273.1 | AAH03273.1 | |||
| BC003993.1 | AAH03993.1 |
More Information
Set Membership: |
|

Genomic Map