Assay Details
Target Gene Details
Entrez Gene ID: | 67204 |
Gene Name: | eukaryotic translation initiation factor 2, subunit 2 (beta) |
Gene Aliases: |
2810026E11Rik, 38kDa, AA408636, AA571381, AA986487, AW822225, D2Ertd303e, EIF2, EIF2B |
Location: |
Chr.2:154871410-154892906 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif2s2 | NM_026030.2 | NP_080306.1 | ||
| AK012338.1 | ||||
| AK012817.1 | BAB28490.2 | |||
| AK076056.1 | ||||
| AK143553.1 | 2 | 4061 | ||
| AK145401.1 | BAE26414.1 | |||
| AK145434.1 | BAE26433.1 | |||
| AK166862.1 | BAE39077.1 | |||
| AK166866.1 | 3 | 3711 | ||
| BC113766.1 | AAI13767.1 |
More Information
Set Membership: |
|

Genomic Map