Assay Details
Target Gene Details
Entrez Gene ID: | 319513 |
Gene Name: | PC-esterase domain containing 1A |
Gene Aliases: |
A930025D01Rik, AI480484, Fam113a |
Location: |
Chr.2:130417684-130424641 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pced1a | NM_001114541.1 | NP_001108013.1 | ||
| NM_178762.4 | NP_848877.2 | |||
| AK031469.1 | 1 | 1768 | BAC27419.1 | |
| AK032354.1 | BAC27830.1 | |||
| AK032762.1 | BAC28010.1 | |||
| AK041939.1 | 1 | 1348 | ||
| AK044590.1 | 1 | 1339 | BAC31990.1 | |
| AK045248.1 | BAC32279.1 | |||
| AK077745.1 | 1 | 1345 | ||
| AK081584.1 | BAC38264.1 | |||
| AK085613.1 | BAC39486.1 | |||
| AK161998.1 | ||||
| AK183067.1 | ||||
| AK183310.1 | ||||
| AK210108.1 | ||||
| BC064802.1 | AAH64802.1 | |||
| BY326318.1 |
More Information
Set Membership: |
|

Genomic Map