Assay Details
Target Gene Details
Entrez Gene ID: | 77799 |
Gene Name: | Src-like-adaptor 2 |
Gene Aliases: |
A930009E21Rik, AI430952, SLAP-2, SLAP2 |
Location: |
Chr.2:156872922-156887241 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sla2 | NM_029983.5 | 2 | 264 | NP_084259.1 |
| AF287467.1 | 2 | 315 | AAL38196.1 | |
| AF434990.1 | 2 | 307 | AAL86403.1 | |
| AK030877.1 | 2 | 265 | BAC27168.1 | |
| AK088672.1 | 2 | 391 | BAC40495.1 | |
| AK138709.1 | 2 | 265 | BAE23754.1 | |
| BC052655.1 | 2 | 363 | AAH52655.1 |
More Information
Set Membership: |
|

Genomic Map