Assay Details
Target Gene Details
Entrez Gene ID: | 20269 |
Gene Name: | sodium channel, voltage-gated, type III, alpha |
Gene Aliases: |
Gm1000, Nav1.3 |
Location: |
Chr.2:65457118-65567527 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scn3a | NM_018732.3 | 24 | 4497 | NP_061202.3 |
| CJ133030.1 | 3 | 182 |
More Information
Set Membership: |
|

Genomic Map