Assay Details
Target Gene Details
Entrez Gene ID: | 545486 |
Gene Name: | tubulin, beta 1 class VI |
Gene Aliases: |
2810484G07Rik, M(beta)1 |
Location: |
Chr.2:174450595-174458380 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tubb1 | NM_001080971.2 | NP_001074440.1 | ||
| BC147322.1 | ||||
| BC147323.1 | ||||
| BC147687.1 | ||||
| BC147698.1 |
More Information
Set Membership: |
|

Genomic Map