Assay Details
Target Gene Details
Entrez Gene ID: | 68527 |
Gene Name: | upper zone of growth plate and cartilage matrix associated |
Gene Aliases: |
1110017I16Rik, AW121955, Grp |
Location: |
Chr.2:4976122-4985748 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ucma | NM_001113558.2 | NP_001107030.1 | ||
| NM_001165932.1 | NP_001159404.1 | |||
| NM_026754.3 | NP_081030.1 | |||
| AK003750.1 | BAB22978.2 | |||
| BC115608.2 | ||||
| BC115609.2 | ||||
| BE648935.1 | ||||
| EF529510.1 | ||||
| EU368184.1 | ||||
| FJ217397.1 | ||||
| FJ217398.1 | ||||
| FJ217399.1 | ||||
| FJ217400.1 |

Genomic Map