Assay Details
Target Gene Details
Entrez Gene ID: | 545527 |
Gene Name: | glutamate rich 6 |
Gene Aliases: |
4932431H17Rik, Fam194a |
Location: |
Chr.3:58616300-58665413 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Erich6 |

Genomic Map