Assay Details
Target Gene Details
Entrez Gene ID: | 229541 |
Gene Name: | DENN/MADD domain containing 4B |
Gene Aliases: |
- |
Location: |
Chr.3:90264920-90280665 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dennd4b | NM_201407.4 | NP_958809.3 | ||
| AK155638.1 | BAE33359.1 | |||
| BY202147.1 |

Genomic Map