Assay Details
Target Gene Details
Entrez Gene ID: | 66292 |
Gene Name: | mitochondrial ribosomal protein S21 |
Gene Aliases: |
1810031B19Rik |
Location: |
Chr.3:95862646-95871604 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps21 | NM_078479.3 | NP_510964.1 | ||
| AB049956.1 | BAB41009.1 | |||
| AK002588.1 | BAC24999.1 | |||
| AK003462.1 | BAC25036.1 | |||
| AK003602.1 | BAC25044.1 | |||
| AK007662.1 | BAC25181.1 | |||
| AK007838.1 | BAC25189.1 | |||
| AK008265.1 | BAC25210.1 | |||
| AK008293.1 | BAC25213.1 | |||
| AK009049.1 | BAC25237.1 | |||
| AK009703.1 | BAC25267.1 | |||
| BC027546.1 | AAH27546.1 | |||
| BC125658.1 | ||||
| BC125660.1 |
More Information
Set Membership: |
|

Genomic Map