Assay Details
Target Gene Details
Entrez Gene ID: | 17829 |
Gene Name: | mucin 1, transmembrane |
Gene Aliases: |
CD227, EMA, Muc-1 |
Location: |
Chr.3:89229056-89233381 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Muc1 | NM_013605.2 | 1 | 61 | NP_038633.1 |
| BC005441.1 | AAH05441.1 | |||
| CJ066040.1 | 1 | 62 | ||
| M84683.1 | AAA39756.1 |

Genomic Map