Assay Details
Target Gene Details
Entrez Gene ID: | 13347 |
Gene Name: | DNA fragmentation factor, alpha subunit |
Gene Aliases: |
A330085O09Rik, DFF35, Dff45, ICAD, ICAD-L, ICAD-S |
Location: |
Chr.4:149104142-149120653 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dffa | NM_001025296.2 | NP_001020467.1 | ||
| NM_010044.3 | NP_034174.2 | |||
| NR_104263.1 | ||||
| AB009375.1 | BAA24140.1 | |||
| AB009376.1 | BAA24141.1 | |||
| AI116789.1 | ||||
| AK039227.1 | BAC30285.1 | |||
| AK051011.1 | BAC34496.1 | |||
| AK079634.1 | BAC37710.1 | |||
| AK154488.1 | ||||
| BC058213.1 | AAH58213.1 | |||
| BI736905.1 |

Genomic Map