Assay Details
Target Gene Details
Entrez Gene ID: | 18477 |
Gene Name: | peroxiredoxin 1 |
Gene Aliases: |
MSP23, NkefA, OSF-3, OSF3, PAG, Paga, PrdxI, PrxI, TDX2, TPxA, Tdpx2, prx1 |
Location: |
Chr.4:116685599-116700000 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prdx1 | NM_011034.4 | NP_035164.1 | ||
| AK002287.1 | BAB21990.1 | |||
| AK008711.1 | BAB25847.1 | |||
| AK010688.1 | BAB27120.1 | |||
| AK083243.1 | BAC38827.1 | |||
| AK150797.1 | BAE29860.1 | |||
| AK151459.1 | BAE30417.1 | |||
| AK151760.1 | BAE30667.1 | |||
| AK163984.1 | ||||
| AK167624.1 | BAE39676.1 | |||
| AK169154.1 | BAE40933.1 | |||
| AK169992.1 | 1 | 1946 | ||
| BC083348.1 | AAH83348.1 | |||
| BC086648.1 | AAH86648.1 | |||
| D16142.1 | BAA03713.1 | |||
| D21252.1 | BAA04796.1 |
More Information
Set Membership: |
|

Genomic Map