Assay Details
Target Gene Details
Entrez Gene ID: | 18028 |
Gene Name: | nuclear factor I/B |
Gene Aliases: |
6720429L07Rik, CTF, E030026I10Rik, NF-I/B, NF1-B, NFI-B |
Location: |
Chr.4:82290173-82505779 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfib | NM_001113209.2 | 9 | 2336 | NP_001106680.1 |
| NM_001113210.2 | 9 | 2336 | NP_001106681.1 | |
| NM_001286127.1 | 9 | 2336 | NP_001273056.1 | |
| NM_001286131.1 | NP_001273060.1 | |||
| NM_008687.6 | NP_032713.3 | |||
| AA500678.1 | 2 | 265 | ||
| AA510173.1 | 1 | 73 | ||
| AK083167.1 | BAC38792.1 | |||
| AK084941.1 | ||||
| AK145267.1 | 8 | 1042 | ||
| AK146480.1 | BAE27203.1 | |||
| AY035852.1 | AAK62034.1 | |||
| AY035853.1 | AAK62035.1 | |||
| BC014290.1 | 9 | 1373 | AAH14290.1 | |
| BC062908.1 | AAH62908.1 | |||
| U57634.1 | AAB49929.1 | |||
| Y07685.1 | 9 | 2268 | CAA68949.1 | |
| Y07686.1 | CAA68950.1 | |||
| Y07687.1 | CAA68951.1 |
More Information
Set Membership: |
|

Genomic Map