Assay Details
Target Gene Details
Entrez Gene ID: | 14745 |
Gene Name: | lysophosphatidic acid receptor 1 |
Gene Aliases: |
AI326300, Edg2, Gpcr26, Kdt2, lpA1, vzg-1 |
Location: |
Chr.4:58435252-58554027 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lpar1 | NM_010336.2 | 2 | 118 | NP_034466.2 |
| NM_172989.1 | 2 | 207 | NP_766577.1 | |
| AK030330.1 | 2 | 170 | BAC26905.1 | |
| BC025425.1 | 2 | 117 | AAH25425.1 | |
| BY716867.1 | 2 | 166 | ||
| CJ152526.1 | 2 | 118 | ||
| CJ175997.1 | 3 | 349 | ||
| U70622.1 | 2 | 159 | AAC52923.1 |
More Information
Set Membership: |
|

Genomic Map