Assay Details
Target Gene Details
Entrez Gene ID: | 18781 |
Gene Name: | phospholipase A2, group IIC |
Gene Aliases: |
- |
Location: |
Chr.4:138725325-138744575 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g2c | NM_008868.3 | 2 | 227 | NP_032894.2 |
| BC029347.1 | 1 | 72 | AAH29347.1 | |
| CA464794.1 | 2 | 230 | ||
| U18119.1 | 1 | 53 | AAC52936.1 |
Target Gene Details
Entrez Gene ID: | 212190 |
Gene Name: | UBX domain protein 10 |
Gene Aliases: |
5730509E04Rik, A830047D02, Ubxd3 |
Location: |
Chr.4:138709837-138738438 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ubxn10 | NM_001285928.1 | NP_001272857.1 | ||
| NM_001285929.1 | NP_001272858.1 | |||
| AK046949.1 | BAC32924.1 |
More Information
Set Membership: |
|

Genomic Map