Assay Details
Target Gene Details
Entrez Gene ID: | 18786 |
Gene Name: | phospholipase A2, activating protein |
Gene Aliases: |
2410007N06, AI536418, AU018445, AW208417, D4Ertd618e, PLA2P, PLAP, Ufd3 |
Location: |
Chr.4:94565139-94603247 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plaa | NM_172695.2 | 6 | 1029 | NP_766283.2 |
| AK019070.1 | 6 | 1035 | ||
| AK075934.1 | 6 | 961 | BAC36064.1 | |
| AK081641.1 | 6 | 920 | ||
| AK153651.1 | ||||
| AK162534.1 | 5 | 3220 | ||
| AK166122.1 | 6 | 934 | ||
| AK186154.1 | 3 | 302 | ||
| BC139355.1 | 6 | 1028 | ||
| BC139356.1 | 6 | 1028 | ||
| BC139773.1 | 6 | 939 |
More Information
Set Membership: |
|

Genomic Map