Assay Details
Target Gene Details
Entrez Gene ID: | 66212 |
Gene Name: | Sec61 beta subunit |
Gene Aliases: |
1190006C12Rik, AI326121, AW122942 |
Location: |
Chr.4:47474661-47483233 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sec61b | NM_024171.2 | NP_077133.1 | ||
| AK004505.1 | BAB23338.1 | |||
| AK008739.1 | BAB25868.1 | |||
| BC002089.1 | AAH02089.1 | |||
| BC081445.1 | AAH81445.1 |
More Information
Set Membership: |
|

Genomic Map