Assay Details
Target Gene Details
Entrez Gene ID: | 269713 |
Gene Name: | CAP-GLY domain containing linker protein 2 |
Gene Aliases: |
B230327O20, CLIP-115, Clip1, Cyln2, WSCR4, mKIAA0291, wbscr4 |
Location: |
Chr.5:134489383-134552468 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clip2 | NM_001039162.2 | 6 | 1617 | NP_001034251.1 |
| NM_009990.3 | 6 | 1617 | NP_034120.2 | |
| AK045967.1 | 6 | 1461 | ||
| AK172928.1 | 6 | 1362 | BAD32206.1 | |
| BC039162.1 | 6 | 1557 | AAH39162.1 | |
| BC053048.1 | 6 | 1617 | AAH53048.1 |
More Information
Set Membership: |
|

Genomic Map