Assay Details
Target Gene Details
Entrez Gene ID: | 330050 |
Gene Name: | family with sequence similarity 185, member A |
Gene Aliases: |
4631428I10, AI847670 |
Location: |
Chr.5:21424861-21488271 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam185a |
Target Gene Details
Entrez Gene ID: | 320118 |
Gene Name: | F-box and leucine-rich repeat protein 13 |
Gene Aliases: |
4921539K22Rik |
Location: |
Chr.5:21483847-21645698 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbxl13 | NM_001199632.1 | 20 | 2298 | NP_001186561.1 |
| NM_177076.4 | 19 | 2199 | NP_796050.2 | |
| AK029566.1 | 19 | 2201 | BAC26515.1 | |
| BC145291.1 | 20 | 2224 | ||
| BC145292.1 | 19 | 2126 | ||
| CO797010.1 | 3 | 318 |

Genomic Map