Assay Details
Target Gene Details
Entrez Gene ID: | 433966 |
Gene Name: | RIKEN cDNA 5730422E09 gene |
Gene Aliases: |
- |
Location: |
Chr.5:149119565-149182036 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 5730422E09Rik | NR_015478.2 |
More Information
Set Membership: |
|

Genomic Map