Assay Details
Target Gene Details
Entrez Gene ID: | 23912 |
Gene Name: | ras homolog gene family, member f |
Gene Aliases: |
AI845056, AV026554, Arhf, Ifld1, Rif |
Location: |
Chr.5:123118180-123132629 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rhof | NM_175092.3 | NP_780301.1 | ||
| AK172212.1 | BAE42885.1 | |||
| BC096597.1 | AAH96597.1 | |||
| DV646753.1 |
More Information
Set Membership: |
|

Genomic Map