Assay Details
Target Gene Details
Entrez Gene ID: | 18979 |
Gene Name: | paraoxonase 1 |
Gene Aliases: |
Pon |
Location: |
Chr.6:5168090-5193987 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pon1 | NM_011134.3 | 4 | 481 | NP_035264.2 |
| BC012706.1 | 4 | 354 | AAH12706.1 | |
| U32684.1 | 4 | 363 | AAC52496.1 | |
| U72636.1 | 4 | 363 | AAB17394.1 |

Genomic Map