Assay Details
Target Gene Details
Entrez Gene ID: | 64177 |
Gene Name: | transient receptor potential cation channel, subfamily V, member 6 |
Gene Aliases: |
CAT, CaT1, Cac, Ecac2, Otrpc3 |
Location: |
Chr.6:41620619-41636480 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trpv6 | NM_022413.4 | 2 | 472 | NP_071858.3 |
| AB037373.1 | 2 | 373 | BAA99538.1 | |
| AJ542487.1 | 2 | 177 | CAD62684.1 | |
| AK077234.1 | 2 | 402 | BAC36699.1 | |
| AK085123.1 | 2 | 448 | ||
| AK168760.1 | 2 | 472 | ||
| BC016101.1 | 2 | 410 | AAH16101.1 |
More Information
Set Membership: |
|

Genomic Map