Assay Details
Target Gene Details
Entrez Gene ID: | 100039953 |
Gene Name: | golgi-associated olfactory signaling regulator |
Gene Aliases: |
Gm2515, Gm581, Goofy |
Location: |
Chr.7:45176349-45179597 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gfy | NM_001195255.1 | NP_001182184.1 | ||
| AB845286.1 | ||||
| BM934591.1 |
Target Gene Details
Entrez Gene ID: | 114640 |
Gene Name: | parathyroid hormone 2 |
Gene Aliases: |
Tifp39, Tip39 |
Location: |
Chr.7:45176565-45181838 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pth2 |

Genomic Map